missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-4-pyranol 97.0+%, TCI America™

Catalog No. T28255G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28255G 5 g
T28251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T28255G Supplier TCI America Supplier No. T28255G
Only null left

Chemical Identifiers

CAS 2081-44-9
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00006633
InChI Key LMYJGUNNJIDROI-UHFFFAOYSA-N
Synonym tetrahydro-2h-pyran-4-ol, tetrahydro-4-pyranol, 4-hydroxytetrahydropyran, tetrahydro-4h-pyran-4-ol, tetrahydro-pyran-4-ol, 2h-pyran-4-ol, tetrahydro, 4-hydroxytetrahydropyan, tetrahydropyran-4-ol, 4-hydroxytetrahydro-4h-pyran, 4-hydroxytetrahydro-2h-pyran
PubChem CID 74956
IUPAC Name oxan-4-ol
SMILES C1COCCC1O

Specifications

Color Colorless
Boiling Point 190°C
Quantity 5 g
Formula Weight 102.13
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetrahydro-4-pyranol