accessibility menu, dialog, popup

UserName

CAS RN 207511-11-3

IUPAC Name: sodium 2-sulfanylidene-2,3-dihydro-1H-1,3-benzodiazole-5-sulfonate dihydrate
Synonyms: sodium 2-sulfanylidene-1,3-dihydro-1,3-benzodiazole-5-sulfonate dihydrate
Molecular Weight (g/mol): 288.27
Molecular Formula: C7H9N2NaO5S2
InChi Key: OIYOMRFNKQEDEH-UHFFFAOYSA-M
SMILES: O.O.[Na+].[O-]S(=O)(=O)C1=CC=C2NC(=S)NC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results