missing translation for 'onlineSavingsMsg'
Learn More

Sodium 2-Mercapto-5-benzimidazolesulfonate Dihydrate 98.0+%, TCI America™

Catalog No. M127025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M127025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M127025G Supplier TCI America Supplier No. M127025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 207511-11-3
Molecular Formula C7H9N2NaO5S2
Molecular Weight (g/mol) 288.27
MDL Number MFCD00192260
InChI Key OIYOMRFNKQEDEH-UHFFFAOYSA-M
Synonym 2-Mercapto-5-benzimidazolesulfonic Acid Sodium Salt
PubChem CID 23676756
IUPAC Name sodium 2-sulfanylidene-2,3-dihydro-1H-1,3-benzodiazole-5-sulfonate dihydrate
SMILES O.O.[Na+].[O-]S(=O)(=O)C1=CC=C2NC(=S)NC2=C1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 252.24
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sodium 2-Mercapto-5-benzimidazolesulfonate Dihydrate
EINECSNumber (5)-0471
TSCA Yes