accessibility menu, dialog, popup

UserName

H3CNCH3NH3CCH3

CAS RN 20734-58-1

IUPAC Name: N1,N1,N8,N8-tetramethylnaphthalene-1,8-diamine
Synonyms: N1,N1,N8,N8-tetramethylnaphthalene-1,8-diamine
Molecular Weight (g/mol): 214.31
Molecular Formula: C14H18N2
InChi Key: GJFNRSDCSTVPCJ-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=CC2=CC=CC(N(C)C)=C12
Boiling Point 
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (5)
  • (2)