missing translation for 'onlineSavingsMsg'
Learn More

1,8-Bis(dimethylamino)naphthalene, 99%

Catalog No. AC169860500
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169860500 50 g Glass bottle
AC169860100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC169860500 Supplier Thermo Scientific Chemicals Supplier No. 169860500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20734-58-1 | C14H18N2 | 214.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20734-58-1
Molecular Formula C14H18N2
Molecular Weight (g/mol) 214.31
MDL Number MFCD00003920
InChI Key GJFNRSDCSTVPCJ-UHFFFAOYSA-N
Synonym 1,8-bis dimethylamino naphthalene, n1,n1,n8,n8-tetramethylnaphthalene-1,8-diamine, proton sponge, 1,8-bis dimethylamino naphtalene, n,n,n',n'-tetramethyl-1,8-naphthalenediamine, proton-sponge, 1,8-naphthalenediamine, n,n,n',n'-tetramethyl, unii-6s79d2p9h8, 1,8-bis-dimethylamino naphthalene, n,n,n',n'-tetramethylnaphthalene-1,8-diamine
PubChem CID 88675
IUPAC Name 1-N,1-N,8-N,8-N-tetramethylnaphthalene-1,8-diamine
SMILES CN(C)C1=CC=CC2=C1C(=CC=C2)N(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 46°C to 50°C
Color Brown-Pink
Boiling Point 144°C to 145°C (5.0 hPa)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 50 g
Fieser 03,22; 04,35; 06,50; 13,293
Merck Index 15, 3432
Solubility Information Solubility in water: insoluble
Formula Weight 214.31
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 1, 8-Bis(dimethylamino)naphthalene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 244-001-6

RUO – Research Use Only