CAS RN 207300-91-2
Sodium 1-Hexanesulfonate Monohydrate, 99%
CAS: 207300-91-2 Molecular Formula: C6H15NaO4S Molecular Weight (g/mol): 206.232 MDL Number: MFCD00149549 InChI Key: CLCGFJYKZGFGSQ-UHFFFAOYSA-M Synonym: 1-hexanesulfonic acid sodium salt monohydrate,sodium hexane-1-sulfonate hydrate,sodium 1-hexanesulfonate monohydrate,n-1-hexanesulfonic acid,sodium 1-hexanesulfonate hydrate,sodium n-hexanesulfonate monohydrate,potassium hexane-1-sulfonate hydrate,sodium hexane-1-sulphonate monohydrate,sodium hexane-1-sulfonate-water 1/1/1,1-hexanesulfonic acid,sodium salt, hydrate 1:1:1 PubChem CID: 23696962 IUPAC Name: sodium;hexane-1-sulfonate;hydrate SMILES: CCCCCCS(=O)(=O)[O-].O.[Na+]
LiChropur™ Sodium 1-Hexanesulfonate monohydrate , ≥99.0% (T), MilliporeSigma™ Supelco™
MDL Number: MFCD00149549 Synonym: 1-Hexanesulfonic acid sodium salt monohydrate
1-Hexanesulfonic Acid Sodium Salt, Monohydrate, 98%, Spectrum™ Chemical
CAS: 207300-91-2