missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ Sodium 1-Hexanesulfonate monohydrate , ≥99.0% (T), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 207300-91-2 |
| Linear Formula | CH3(CH2)4 CH2 SO3 Na · H2O |
| Quantity | 50 g |
| Molecular Weight (g/mol) | 206.24 |
| Formula Weight | 206.24 |
| Percent Purity | ≥99.0% (T) |
| Grade | For ion pair chromatography |
| Physical Form | Solid |
| Product Line | LiChropur |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany