accessibility menu, dialog, popup

UserName

CH2NOH

CAS RN 207129-36-0

IUPAC Name: {5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl}methanol
Synonyms: {5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl}methanol
Molecular Weight (g/mol): 167.25
Molecular Formula: C10H17NO
InChi Key: GAFZBOMPQVRGKU-UHFFFAOYNA-N
SMILES: OCC1CC2CCN1CC2C=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results