CAS RN 20662-53-7
IUPAC Name:
1-(piperidin-4-yl)-2,3-dihydro-1H-1,3-benzodiazol-2-one
Synonyms:
1-(piperidin-4-yl)-3H-1,3-benzodiazol-2-one
Molecular Weight (g/mol):
217.27
Molecular Formula:
C12H15N3O
InChi Key:
BYNBAMHAURJNTR-UHFFFAOYSA-N
SMILES:
O=C1NC2=CC=CC=C2N1C1CCNCC1
1
–
2
of
2
results
1
Promotions Available
4-(2-Keto-1-benzimidazolinyl)piperidine, 97%
4-(2-Keto-1-benzimidazolinyl)piperidine, 97%
CAS: 20662-53-7 Molecular Formula: C12H15N3O Molecular Weight (g/mol): 217.27 MDL Number: MFCD00005714 InChI Key: BYNBAMHAURJNTR-UHFFFAOYSA-N Synonym: 4-2-keto-1-benzimidazolinyl piperidine,1-piperidin-4-yl-1h-benzo d imidazol-2 3h-one,4-2-keto-1-benzimidozolinyl piperidine,unii-4rp0aed2pc,1-piperidin-4-yl-1h-benzimidazol-2-ol,4rp0aed2pc,1-4-piperidinyl-1,3-dihydro-2h-benzimidazol-2-one,1-piperidin-4-yl-2,3-dihydro-1h-1,3-benzodiazol-2-one,1-piperidin-4-yl-3h-1,3-benzodiazol-2-one,1,3-dihydro-1-4-piperidinyl-2h-benzimidazol-2-one PubChem CID: 88638 SMILES: O=C1NC2=CC=CC=C2N1C1CCNCC1
2
Promotions Available
4-(2-Keto-1-benzimidazolinyl)piperidine, 97%, Thermo Scientific™