accessibility menu, dialog, popup

UserName

CAS RN 2058-46-0

ClH3CNCH3(S)(R)(S)OH(R)OHOH(S)CH3HOO(S)OHOH2NOHOH(Z)

CAS RN 2058-46-0

IUPAC Name: hydrogen (2Z,4S,4aR,5S,5aR,6S,12aS)-2-[amino(hydroxy)methylidene]-4-(dimethylamino)-5,6,10,11,12a-pentahydroxy-6-methyl-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride
Synonyms: hydrogen biostat chloride
Molecular Weight (g/mol): 496.9
Molecular Formula: C22H25ClN2O9
InChi Key: IZRXCFCWDMSBQY-NXURVHCDSA-N
SMILES: [H+].[Cl-].CN(C)[C@H]1[C@@H]2[C@@H](O)[C@H]3C(=C(O)C4=C(O)C=CC=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (4)
Color 
  • (2)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)