missing translation for 'onlineSavingsMsg'
Learn More

Oxytetracycline Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012179
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012179 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012179 Supplier Merck Emd Millipore Supplier No. PHR1325500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Oxytetracycline is a broad-spectrum antibiotic drug widely employed in veterinary applications, particularly for the management of infections in pig productions. It acts by inhibiting the protein synthesis in gram-positive and gram-negative bacteria.

Specifications

CAS 2058-46-0
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C22H24N2O9 · HCl
MDL Number MFCD00135815
Synonym Oxytetracycline hydrochloride; 5-Hydroxytetracycline hydrochloride
Type Certified Reference Material
Quantity 500 mg
Formula Weight 496.89
Grade Certified Reference Material