missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oxytetracycline Hydrochloride, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| Chemical Name or Material | Pharmacopeia & Metrological Institutes Standards |
| CAS | 2058-46-0 |
| Flash Point | Not Applicable |
| Molecular Formula | C22H24N2O9 · HCl |
| Linear Formula | C22H24N2O9 · HCl |
| Quantity | 20 mg |
| MDL Number | MFCD00135815 |
| Synonym | 5-Hydroxytetracycline hydrochloride |
| Molecular Weight (g/mol) | 496.89 |
| Formula Weight | 496.89 |
| Show More |