accessibility menu, dialog, popup

UserName

CAS RN 20570-96-1

IUPAC Name: dihydrogen benzylhydrazine dichloride
Synonyms: dihydrogen benzylhydrazine dichloride
Molecular Weight (g/mol): 195.09
Molecular Formula: C7H12Cl2N2
InChi Key: MSJHOJKVMMEMNX-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NNCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (1)
  • (2)
  • (3)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (3)
  • (3)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)