missing translation for 'onlineSavingsMsg'
Learn More

Benzylhydrazine Dihydrochloride 98.0+%, TCI America™

Catalog No. B34715G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B34715G Supplier TCI America Supplier No. B34715G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20570-96-1
Molecular Formula C7H12Cl2N2
Molecular Weight (g/mol) 195.087
MDL Number MFCD00012921
InChI Key MSJHOJKVMMEMNX-UHFFFAOYSA-N
Synonym benzylhydrazine dihydrochloride, benzylhydrazine.2hcl, 1-benzylhydrazine dihydrochloride, benzyl-hydrazine dihydrochloride, hydrazine, phenylmethyl-, dihydrochloride, hydrazine, benzyl-, dihydrochloride, phenylmethyl hydrazine dihydrochloride, benzyl hydrazine di hcl, benzyl-hydrazine hydrochloride, benzylhydrazinedihydrochloride
PubChem CID 146540
IUPAC Name benzylhydrazine;dihydrochloride
SMILES C1=CC=C(C=C1)CNN.Cl.Cl

Specifications

Melting Point 145°C
Color White-Yellow
Quantity 5 g
UN Number 2811
Formula Weight 195.09
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzylhydrazine Dihydrochloride
EINECSNumber (3)-0466
RTECSNumber MU8540000
TSCA No