accessibility menu, dialog, popup

UserName

CAS RN 20555-91-3

IUPAC Name: 1,2-dichloro-4-iodobenzene
Synonyms: 1,2-dichloro-4-iodobenzene
Molecular Weight (g/mol): 272.89
Molecular Formula: C6H3Cl2I
InChi Key: NADPFZNWCQIJJW-UHFFFAOYSA-N
SMILES: ClC1=CC=C(I)C=C1Cl
Percent Purity 
  • (1)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (5)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)