missing translation for 'onlineSavingsMsg'
Learn More

1,2-dichloro-4-iodobenzene, Thermo Scientific™

Catalog No. p-4391294
Change view
Click to view available options
Quantity:
10g
25g
100g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB06155EA 10g
BTB06155EB 25g
BTB06155FA 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BTB06155EA Supplier Thermo Scientific Chemicals Supplier No. BTB06155EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20555-91-3
Molecular Formula C6H3Cl2I
Molecular Weight (g/mol) 272.894
InChI Key NADPFZNWCQIJJW-UHFFFAOYSA-N
Synonym 3,4-dichloroiodobenzene, benzene, 1,2-dichloro-4-iodo, maybridge1_001437, pubchem3691, acmc-209fbv, 3,4-dichloro-iodobenzene, 1,2-dichloro-5-iodobenzene, 1-iodo-3,4-dichlorobenzene, 3,4-dichloro-1-iodobenzene, 4-iodo-1,2-dichlorobenzene
PubChem CID 88591
IUPAC Name 1,2-dichloro-4-iodobenzene
SMILES C1=CC(=C(C=C1I)Cl)Cl

Specifications

Assay Percent Range 97%
Linear Formula IC6H3Cl2
Quantity 10g
Formula Weight 272.9
Percent Purity 97%
Chemical Name or Material 1, 2-Dichloro-4-iodobenzene