missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloro-4-iodobenzene, 98%

Catalog No. AAA1344018
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1344018 50 g
AAA1344009 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1344018 Supplier Thermo Scientific Chemicals Supplier No. A1344018
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 20555-91-3 | C6H3Cl2I | 272.894 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20555-91-3
Molecular Formula C6H3Cl2I
Molecular Weight (g/mol) 272.894
MDL Number MFCD00019014
InChI Key NADPFZNWCQIJJW-UHFFFAOYSA-N
Synonym 3,4-dichloroiodobenzene, benzene, 1,2-dichloro-4-iodo, maybridge1_001437, pubchem3691, acmc-209fbv, 3,4-dichloro-iodobenzene, 1,2-dichloro-5-iodobenzene, 1-iodo-3,4-dichlorobenzene, 3,4-dichloro-1-iodobenzene, 4-iodo-1,2-dichlorobenzene
PubChem CID 88591
IUPAC Name 1,2-dichloro-4-iodobenzene
SMILES C1=CC(=C(C=C1I)Cl)Cl

Specifications

Melting Point 29°C to 31°C
Boiling Point 116°C to 118°C (12 mmHg)
Refractive Index 1.648
Quantity 50 g
Beilstein 2245136
Sensitivity Light sensitive
Formula Weight 272.9
Percent Purity 98%
Chemical Name or Material 1,2-Dichloro-4-iodobenzene
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 243-874-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only