missing translation for 'onlineSavingsMsg'
Learn More

1,2-Dichloro-4-iodobenzene 98.0+%, TCI America™

Catalog No. D143610G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D143610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D143610G Supplier TCI America Supplier No. D143610G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20555-91-3
Molecular Formula C6H3Cl2I
Molecular Weight (g/mol) 272.894
MDL Number MFCD00019014
InChI Key NADPFZNWCQIJJW-UHFFFAOYSA-N
Synonym 3,4-dichloroiodobenzene, benzene, 1,2-dichloro-4-iodo, maybridge1_001437, pubchem3691, acmc-209fbv, 3,4-dichloro-iodobenzene, 1,2-dichloro-5-iodobenzene, 1-iodo-3,4-dichlorobenzene, 3,4-dichloro-1-iodobenzene, 4-iodo-1,2-dichlorobenzene
PubChem CID 88591
IUPAC Name 1,2-dichloro-4-iodobenzene
SMILES C1=CC(=C(C=C1I)Cl)Cl

Specifications

Quantity 10 g
Formula Weight 272.89
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1,2-Dichloro-4-iodobenzene