accessibility menu, dialog, popup

UserName

CAS RN 2049-94-7

H3CCH3

CAS RN 2049-94-7

IUPAC Name: (3-methylbutyl)benzene
Synonyms: benzene, (3-methylbutyl)-
Molecular Weight (g/mol): 148.25
Molecular Formula: C11H16
InChi Key: XNXIYYFOYIUJIW-UHFFFAOYSA-N
SMILES: CC(C)CCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results