accessibility menu, dialog, popup

UserName

H3COO(R)OCH3CH3(R)HNCH3O(S)H2NOHPOHOHO

CAS RN 204255-11-8

IUPAC Name: ethyl (3R,4R,5S)-5-amino-4-acetamido-3-(pentan-3-yloxy)cyclohex-1-ene-1-carboxylate; phosphoric acid
Synonyms: tamiflu
Molecular Weight (g/mol): 410.4
Molecular Formula: C16H31N2O8P
InChi Key: PGZUMBJQJWIWGJ-ONAKXNSWSA-N
SMILES: OP(O)(O)=O.CCOC(=O)C1=C[C@@H](OC(CC)CC)[C@H](NC(C)=O)[C@@H](N)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results