missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oseltamivir phosphate, 98%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White to Off-White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (HPLC) |
| Quantity | 5 g |
| Formula Weight | 410.4 |
| Percent Purity | 98% |
| Chemical Name or Material | Oseltamivir phosphate |
Safety and Handling
| Hazard Category | Serious eye damage/eye irritation (category 2), Acute toxicity (category 4), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2) |
| Hazard Statement | Causes serious eye irritation, Harmful if swallowed, May cause respiratory irritation, Causes skin irritation |
RUO – Research Use Only