missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Oseltamivir phosphate
Description
Oseltamivir phosphate is a viral neuraminidase inhibitor prodrug; metabolized to GS 4071. Metabolite exhibits potent inhibition of neuraminidase from multiple influenza strains (IC50 values range from 0.3 to 2 nM). Inhibits cellular effects of influenza infection in vitro (IC50 values range from 0.6 to 150 nM). Displays highest potency against H1N1 and H3N2 influenza A strains. Inhibits H1N1 influenza infection in mice.
Specifications
Specifications
| Chemical Name or Material | Ethyl (3 R,4 R,5 S)-4-Acetamido-5-amino-3-(pentan-3-yloxy)cyclohex-1-ene-1-carboxylate phosphate |
| CAS | 204255-11-8 |
| Quantity | 100 mg |
| Target | Hydrolase Inhibitors |
| Molecular Formula | C16H28N2O4.H3PO4 |
| Purity | 0.98 |