accessibility menu, dialog, popup

UserName

CAS RN 2041-15-8

OHOHHO

CAS RN 2041-15-8

IUPAC Name: cyclohexane-1,3,5-triol
Synonyms: cyclohexane-1,3,5-triol
Molecular Weight (g/mol): 132.16
Molecular Formula: C6H12O3
InChi Key: FSDSKERRNURGGO-UHFFFAOYSA-N
SMILES: OC1CC(O)CC(O)C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results