accessibility menu, dialog, popup

UserName

BrCH2

CAS RN 2039-86-3

IUPAC Name: 1-bromo-3-ethenylbenzene
Synonyms: 1-bromo-3-ethenylbenzene
Molecular Weight (g/mol): 183.05
Molecular Formula: C8H7Br
InChi Key: KQJQPCJDKBKSLV-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(C=C)=C1
Boiling Point 
  • (3)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (1)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (5)
  • (1)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (3)