missing translation for 'onlineSavingsMsg'
Learn More

3-Bromostyrene (stabilized with TBC) 97.0+%, TCI America™

Catalog No. B18975G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18975G 5 g
B18971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18975G Supplier TCI America Supplier No. B18975G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2039-86-3
Molecular Formula C8H7Br
Molecular Weight (g/mol) 183.048
MDL Number MFCD00000088
InChI Key KQJQPCJDKBKSLV-UHFFFAOYSA-N
Synonym 3-bromostyrene, 1-bromo-3-vinylbenzene, m-bromostyrene, benzene, 1-bromo-3-ethenyl, 1-bromo-3-vinyl-benzene, unii-6xe3sf241y, 3-bromo-styrene, pubchem23859, acmc-1cmdc, 1-bromo-3-vinylbenzene #
PubChem CID 74870
IUPAC Name 1-bromo-3-ethenylbenzene
SMILES C=CC1=CC(=CC=C1)Br

Specifications

Color Yellow
Boiling Point 94°C
Quantity 5 g
Formula Weight 183.05
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromostyrene (stabilized with TBC)
TSCA No
Recommended Storage Refrigerator