accessibility menu, dialog, popup

UserName

CAS RN 2039-67-0

IUPAC Name: hydrogen 2-(3-methoxyphenyl)ethan-1-amine chloride
Synonyms: hydrogen 2-(3-methoxyphenyl)ethanamine chloride
Molecular Weight (g/mol): 187.67
Molecular Formula: C9H14ClNO
InChi Key: KDRBQDYHPKRFGX-UHFFFAOYSA-N
SMILES: [H+].[Cl-].COC1=CC=CC(CCN)=C1
Boiling Point 
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (7)
  • (1)
  • (3)
  • (3)