CAS RN 20265-37-6
IUPAC Name:
3-methoxy-2-nitropyridine
Synonyms:
3-methoxy-2-nitropyridine
Molecular Weight (g/mol):
154.13
Molecular Formula:
C6H6N2O3
InChi Key:
LSXHCFSGOBFNDX-UHFFFAOYSA-N
SMILES:
COC1=C(N=CC=C1)[N+]([O-])=O
1
–
1
of
1
results
1
3-Methoxy-2-nitropyridine 98.0+%, TCI America™
CAS: 20265-37-6 Molecular Formula: C6H6N2O3 Molecular Weight (g/mol): 154.125 MDL Number: MFCD00222276 InChI Key: LSXHCFSGOBFNDX-UHFFFAOYSA-N Synonym: 2-nitro-3-methoxypyridine,3-methoxy-2-nitro-pyridine,3-methoxy-2-nitropyridin,pyridine, 3-methoxy-2-nitro,pubchem3990,2-nitro-2-methoxypyridine,acmc-209f7b,3-methoxy-2-nitro pyridine,3m2np,ksc494o8l PubChem CID: 556063 IUPAC Name: 3-methoxy-2-nitropyridine SMILES: COC1=C(N=CC=C1)[N+](=O)[O-]