missing translation for 'onlineSavingsMsg'
Learn More

3-Methoxy-2-nitropyridine 98.0+%, TCI America™

Catalog No. M135925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M135925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M135925G Supplier TCI America Supplier No. M135925G
Only null left

Chemical Identifiers

CAS 20265-37-6
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.125
MDL Number MFCD00222276
InChI Key LSXHCFSGOBFNDX-UHFFFAOYSA-N
Synonym 2-nitro-3-methoxypyridine, 3-methoxy-2-nitro-pyridine, 3-methoxy-2-nitropyridin, pyridine, 3-methoxy-2-nitro, pubchem3990, 2-nitro-2-methoxypyridine, acmc-209f7b, 3-methoxy-2-nitro pyridine, 3m2np, ksc494o8l
PubChem CID 556063
IUPAC Name 3-methoxy-2-nitropyridine
SMILES COC1=C(N=CC=C1)[N+](=O)[O-]

Specifications

Melting Point 74°C
Color Yellow
Quantity 25 g
Formula Weight 154.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Methoxy-2-nitropyridine