accessibility menu, dialog, popup

UserName

CAS RN 2023-48-5

IUPAC Name: difluorotriphenylbismuthane
Synonyms: difluorotriphenylbismuthane
Molecular Weight (g/mol): 478.3
Molecular Formula: C18H15BiF2
InChi Key: OSGJIYNQSIPLAQ-UHFFFAOYSA-L
SMILES: F[Bi](F)(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results