CAS RN 20187-55-7
IUPAC Name:
sodium 2-[(1-benzyl-1H-indazol-3-yl)oxy]acetate
Synonyms:
bendazac
Molecular Weight (g/mol):
304.28
Molecular Formula:
C16H13N2NaO3
InChi Key:
GHFGHIKJGVMEFT-UHFFFAOYSA-M
SMILES:
[Na+].[O-]C(=O)COC1=NN(CC2=CC=CC=C2)C2=CC=CC=C12
1
–
2
of
2
results
1
Bendazac, Thermo Scientific Chemicals
CAS: 20187-55-7 Molecular Formula: C16H13N2NaO3 Molecular Weight (g/mol): 304.28 InChI Key: GHFGHIKJGVMEFT-UHFFFAOYSA-M IUPAC Name: sodium 2-[(1-benzyl-1H-indazol-3-yl)oxy]acetate SMILES: [Na+].[O-]C(=O)COC1=NN(CC2=CC=CC=C2)C2=CC=CC=C12
2
Bendazac 97.0+%, TCI America™
CAS: 20187-55-7 Molecular Formula: C16H14N2O3 Molecular Weight (g/mol): 282.299 MDL Number: MFCD00866158 InChI Key: BYFMCKSPFYVMOU-UHFFFAOYSA-N Synonym: [(1-Benzyl-3-indazolyl)oxy]acetic Acid, 1-Benzyl-3-[1-(carboxy)methoxy]indazole PubChem CID: 2313 ChEBI: CHEBI:31257 IUPAC Name: 2-(1-benzylindazol-3-yl)oxyacetic acid SMILES: C1=CC=C(C=C1)CN2C3=CC=CC=C3C(=N2)OCC(=O)O