missing translation for 'onlineSavingsMsg'
Learn More

Bendazac, Thermo Scientific Chemicals

Catalog No. AC464920050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464920050 5 g
AC464920010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464920050 Supplier Thermo Scientific Chemicals Supplier No. 464920050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 20187-55-7
Molecular Formula C16H13N2NaO3
Molecular Weight (g/mol) 304.28
InChI Key GHFGHIKJGVMEFT-UHFFFAOYSA-M
IUPAC Name sodium 2-[(1-benzyl-1H-indazol-3-yl)oxy]acetate
SMILES [Na+].[O-]C(=O)COC1=NN(CC2=CC=CC=C2)C2=CC=CC=C12

Specifications

Melting Point 160.0°C
Quantity 5 g
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in chloroform, ethanol, acetone
Formula Weight 282.3
Chemical Name or Material Bendazac
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

EINECSNumber 243-569-2

RUO – Research Use Only