accessibility menu, dialog, popup

UserName

CAS RN 20126-76-5

IUPAC Name: (1R)-4-methyl-1-(propan-2-yl)cyclohex-3-en-1-ol
Synonyms: (1R)-1-isopropyl-4-methylcyclohex-3-en-1-ol
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: WRYLYDPHFGVWKC-JTQLQIEISA-N
SMILES: CC(C)[C@@]1(O)CCC(C)=CC1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)