missing translation for 'onlineSavingsMsg'
Learn More

(-)-Terpinen-4-ol 95.0+%, TCI America™

Catalog No. M031925ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M031925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M031925ML Supplier TCI America Supplier No. M031925ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20126-76-5
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.253
MDL Number MFCD00167108
InChI Key WRYLYDPHFGVWKC-JTQLQIEISA-N
Synonym --terpinen-4-ol, --4-terpineol, unii-8vi196vs5t, p-menth-1-en-4-ol, r--, r-p-menth-1-en-4-ol, 3-cyclohexen-1-ol, 4-methyl-1-1-methylethyl-, r, 3-cyclohexen-1-ol, 4-methyl-1-1-methylethyl-, 1r, r-1-isopropyl-4-methyl-3-cyclohexen-1-ol, l-4-terpineneol, l-4-terpineol
PubChem CID 5325830
IUPAC Name (1R)-4-methyl-1-propan-2-ylcyclohex-3-en-1-ol
SMILES CC1=CCC(CC1)(C(C)C)O

Specifications

Color Yellow
Boiling Point 210°C
Quantity 25 mL
Formula Weight 154.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (-)-Terpinen-4-ol
EINECSNumber (3)-2323&(3)-2331
TSCA Yes