accessibility menu, dialog, popup

UserName

H3COONO

CAS RN 20120-24-5

IUPAC Name: ethyl 3-(morpholin-4-yl)propanoate
Synonyms: ethyl 3-(morpholin-4-yl)propanoate
Molecular Weight (g/mol): 187.24
Molecular Formula: C9H17NO3
InChi Key: WRBIQTVRBWJCQT-UHFFFAOYSA-N
SMILES: CCOC(=O)CCN1CCOCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results