missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(Morpholino)propionate 97.0+%, TCI America™

Catalog No. E07845G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07845G 5 g
E07841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07845G Supplier TCI America Supplier No. E07845G

Chemical Identifiers

CAS 20120-24-5
Molecular Formula C9H17NO3
Molecular Weight (g/mol) 187.24
MDL Number MFCD00014622
InChI Key WRBIQTVRBWJCQT-UHFFFAOYSA-N
PubChem CID 88374
IUPAC Name ethyl 3-(morpholin-4-yl)propanoate
SMILES CCOC(=O)CCN1CCOCC1

Specifications

Color Yellow
Boiling Point 118°C
Quantity 5 g
Synonym ethyl 4-morpholinepropionate, ethyl 3-morpholino propionate, ethyl 3-morpholinopropanoate, ethyl 3-4-morpholino propionate, 3-morpholin-4-yl propionic acid ethyl ester, acmc-209f5y, ethyl 3-morpholinopropionate, ethyl-3-morpholinoproprionate, ethyl3-morpholino propionate, tos-bb-1256
Formula Weight 187.24
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Ethyl 3-(Morpholino)propionate