accessibility menu, dialog, popup

UserName

CAS RN 19983-20-1

IUPAC Name: 1-(thiophen-2-yl)piperidine
Synonyms: 1-(thiophen-2-yl)piperidine
Molecular Weight (g/mol): 167.27
Molecular Formula: C9H13NS
InChi Key: SVUBKHLEMUNAQT-UHFFFAOYSA-N
SMILES: C1CCN(CC1)C1=CC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results