accessibility menu, dialog, popup

UserName

CH3NNH3CH3CCH3ClCH3H3CCl(E/Z)(E/Z)(E/Z)

CAS RN 199444-11-6

IUPAC Name: 2-(2-{2-chloro-3-[2-(1,3,3-trimethyl-2,3-dihydro-1H-indol-2-ylidene)ethylidene]cyclohex-1-en-1-yl}ethenyl)-1,3,3-trimethyl-3H-indol-1-ium chloride
Synonyms: 2-(2-{2-chloro-3-[2-(1,3,3-trimethylindol-2-ylidene)ethylidene]cyclohex-1-en-1-yl}ethenyl)-1,3,3-trimethylindol-1-ium chloride
Molecular Weight (g/mol): 519.55
Molecular Formula: C32H36Cl2N2
InChi Key: BPSIJFMUSNMMAL-UHFFFAOYSA-M
SMILES: [Cl-].CN1C2=CC=CC=C2C(C)(C)C1=CC=C1CCCC(C=CC2=[N+](C)C3=CC=CC=C3C2(C)C)=C1Cl
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)