accessibility menu, dialog, popup

UserName

CH3SOONHN

CAS RN 19816-88-7

IUPAC Name: N'-(1,3-diphenylpropan-2-ylidene)-4-methylbenzene-1-sulfonohydrazide
Synonyms: N'-(1,3-diphenylpropan-2-ylidene)-4-methylbenzenesulfonohydrazide
Molecular Weight (g/mol): 378.49
Molecular Formula: C22H22N2O2S
InChi Key: GDXUEUWCUUAZFM-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)NN=C(CC1=CC=CC=C1)CC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results