accessibility menu, dialog, popup

UserName

CAS RN 19781-73-8

H3C

CAS RN 19781-73-8

IUPAC Name: heptadecylcyclohexane
Synonyms: heptadecylcyclohexane
Molecular Weight (g/mol): 322.62
Molecular Formula: C23H46
InChi Key: VLHZQNGNJQYAIZ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC1CCCCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results