missing translation for 'onlineSavingsMsg'
Learn More

Heptadecylcyclohexane 97.0+%, TCI America™

Catalog No. H04435G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H04435G Supplier TCI America Supplier No. H04435G
Only null left

Chemical Identifiers

CAS 19781-73-8
Molecular Formula C23H46
Molecular Weight (g/mol) 322.621
MDL Number MFCD00051453
InChI Key VLHZQNGNJQYAIZ-UHFFFAOYSA-N
Synonym 1-Cyclohexylheptadecane
PubChem CID 29777
IUPAC Name heptadecylcyclohexane
SMILES CCCCCCCCCCCCCCCCCC1CCCCC1

Specifications

Melting Point 36°C
Color White-Yellow
Boiling Point 397°C
Quantity 5 g
Formula Weight 322.62
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Heptadecylcyclohexane
TSCA No
Recommended Storage Refrigerator