accessibility menu, dialog, popup

UserName

H3COCH3

CAS RN 19780-10-0

IUPAC Name: dodecan-5-one
Synonyms: dodecan-5-one
Molecular Weight (g/mol): 184.32
Molecular Formula: C12H24O
InChi Key: DOXYUCZSYSEALW-UHFFFAOYSA-N
SMILES: CCCCCCCC(=O)CCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results