missing translation for 'onlineSavingsMsg'
Learn More

5-Dodecanone 97.0+%, TCI America™

Catalog No. D15365ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D15365ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D15365ML Supplier TCI America Supplier No. D15365ML
Only null left

Chemical Identifiers

CAS 19780-10-0
Molecular Formula C12H24O
Molecular Weight (g/mol) 184.32
MDL Number MFCD00015290
InChI Key DOXYUCZSYSEALW-UHFFFAOYSA-N
Synonym Butyl Heptyl Ketone
PubChem CID 29771
IUPAC Name dodecan-5-one
SMILES CCCCCCCC(=O)CCCC

Specifications

Color Yellow
Quantity 5 mL
Formula Weight 184.32
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Dodecanone