accessibility menu, dialog, popup

UserName

OHHS

CAS RN 19721-22-3

IUPAC Name: 3-sulfanylpropan-1-ol
Synonyms: 1-propanol, 3-mercapto-
Molecular Weight (g/mol): 92.16
Molecular Formula: C3H8OS
InChi Key: SHLSSLVZXJBVHE-UHFFFAOYSA-N
SMILES: OCCCS
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)