missing translation for 'onlineSavingsMsg'
Learn More

3-Mercapto-1-propanol 97.0+%, TCI America™

Catalog No. M12065G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12065G 5 g
M120625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12065G Supplier TCI America Supplier No. M12065G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 19721-22-3
Molecular Formula C3H8OS
Molecular Weight (g/mol) 92.156
MDL Number MFCD00192262
InChI Key SHLSSLVZXJBVHE-UHFFFAOYSA-N
Synonym 3-mercapto-1-propanol, 1-propanol, 3-mercapto, 3-mercaptopropanol, 3-hydroxypropanethiol, 3-hydroxypropyl mercaptan, 3-mecapto-1-propanol, 3-mecaptopropan-1-ol, 1-mercapto-3-propanol, 3-hydroxypropylmercaptan, 3-mercaptopropan-1-ol
PubChem CID 88211
IUPAC Name 3-sulfanylpropan-1-ol
SMILES C(CO)CS

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
UN Number 2810
Formula Weight 92.16
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Mercapto-1-propanol
RTECSNumber UB7630000
TSCA Yes