CAS RN 19686-73-8
IUPAC Name:
1-bromopropan-2-ol
Synonyms:
2-propanol, 1-bromo- C3H7BrO
Molecular Weight (g/mol):
138.99
Molecular Formula:
C3H7BrO
InChi Key:
WEGOLYBUWCMMMY-UHFFFAOYNA-N
SMILES:
CC(O)CBr
1
–
1
of
1
results
1
1-Bromo-2-propanol (contains ca. 20% 2-Bromo-1-propanol) (stabilized with MgO) 75.0+%, TCI America™
CAS: 19686-73-8 Molecular Formula: C3H7BrO Molecular Weight (g/mol): 138.992 MDL Number: MFCD00004529 InChI Key: WEGOLYBUWCMMMY-UHFFFAOYSA-N Synonym: 1-bromo-2-propanol,2-propanol, 1-bromo,propylene bromohydrin,2-hydroxypropyl bromide,1-bromopropanol-2,ccris 5978,1-bromo-2-hydroxypropane,2-hydroxybromopropane,1-bromo-propan-2-ol,pubchem12555 PubChem CID: 29740 IUPAC Name: 1-bromopropan-2-ol SMILES: CC(CBr)O