missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-2-propanol (contains ca. 20% 2-Bromo-1-propanol) (stabilized with MgO) 75.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 25 g |
| UN Number | 2920 |
| Formula Weight | 138.99 |
| Percent Purity | ≥75.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 1-Bromo-2-propanol (contains ca. 20% 2-Bromo-1-propanol) (stabilized with MgO) |
Safety and Handling
| EINECSNumber | (2)-2002 |
| RTECSNumber | UA7388000 |
| TSCA | Yes |
| Recommended Storage | Refrigerator |