accessibility menu, dialog, popup

UserName

CAS RN 19578-68-8

IUPAC Name: 1-(benzyloxy)-4-iodobenzene
Synonyms: 1-(benzyloxy)-4-iodobenzene
Molecular Weight (g/mol): 310.13
Molecular Formula: C13H11IO
InChi Key: MPWFGAWFTAZWKZ-UHFFFAOYSA-N
SMILES: IC1=CC=C(OCC2=CC=CC=C2)C=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)