missing translation for 'onlineSavingsMsg'
Learn More

1-Benzyloxy-4-iodobenzene, 98+%

Catalog No. AAL1609214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1609214 25 g
AAL1609206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1609214 Supplier Thermo Scientific Chemicals Supplier No. L1609214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 19578-68-8 | C13H11IO | 310.134 g/mol

1-Benzyloxy-4-iodobenzene is used as an emulgator and as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Benzyloxy-4-iodobenzene is used as an emulgator and as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to light. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 19578-68-8
Molecular Formula C13H11IO
Molecular Weight (g/mol) 310.134
MDL Number MFCD00079704
InChI Key MPWFGAWFTAZWKZ-UHFFFAOYSA-N
PubChem CID 519671
IUPAC Name 1-iodo-4-phenylmethoxybenzene
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)I

Specifications

Melting Point 60°C to 64°C
Boiling Point 140°C (1 mmHg)
Quantity 25 g
Beilstein 3051495
Sensitivity Light sensitive
Synonym 1-benzyloxy-4-iodobenzene, 4-benzyloxyiodobenzene, 4-iodobenzyloxybenzene, benzyl 4-iodophenyl ether, benzene, 1-iodo-4-phenylmethoxy, 1-benzyloxy-4-iodo-benzene, benzene,1-iodo-4-phenylmethoxy, acmc-20amon, 4-benzyloxy iodobenzene
Solubility Information Insoluble in water.
Formula Weight 310.14
Percent Purity ≥98%
Chemical Name or Material 1-Benzyloxy-4-iodobenzene
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only