accessibility menu, dialog, popup

UserName

CAS RN 19438-10-9

IUPAC Name: methyl 3-hydroxybenzoate
Synonyms: methyl M-hydroxybenzoate
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: YKUCHDXIBAQWSF-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(O)=C1
Boiling Point 
  • (3)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (1)
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (3)
  • (2)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)