missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Hydroxybenzoate 98.0+%, TCI America™

Catalog No. H0215500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0215500G 500 g
H021525G 25 g
H0215100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0215500G Supplier TCI America Supplier No. H0215500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19438-10-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002295
InChI Key YKUCHDXIBAQWSF-UHFFFAOYSA-N
Synonym methyl m-hydroxybenzoate, 3-hydroxybenzoic acid methyl ester, m-carbomethoxyphenol, methyl m-oxybenzoate, m-hydroxybenzoic acid methyl ester, benzoic acid, 3-hydroxy-, methyl ester, 3-hydroxy-benzoic acid methyl ester, unii-569m9g3stj, 3-methoxycarbonyl phenol, benzoic acid, m-hydroxy-, methyl ester
PubChem CID 88068
IUPAC Name methyl 3-hydroxybenzoate
SMILES COC(=O)C1=CC=CC(O)=C1

Specifications

Melting Point 73°C
Color White
Boiling Point 178°C
Quantity 500 g
Formula Weight 152.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3-Hydroxybenzoate
EINECSNumber (3)-1585
TSCA No